Merck
  • Atrial natriuretic peptide inhibits lipopolysaccharide-induced acute lung injury.
Atrial natriuretic peptide inhibits lipopolysaccharide-induced acute lung injury.
Pulmonary pharmacology & therapeutics (2014-01-28)
Takashi Nojiri, Hiroshi Hosoda, Takeshi Tokudome, Koichi Miura, Shin Ishikane, Toru Kimura, Yasushi Shintani, Masayoshi Inoue, Noriyoshi Sawabata, Mikiya Miyazato, Meinoshin Okumura, Kenji Kangawa
ABSTRACT
We recently reported that administration of atrial natriuretic peptide during the perioperative period has prophylactic effects with respect to not only cardiovascular but also respiratory complications following pulmonary resection. However, its mechanisms are not well understood. The objective of the present study was to investigate the mechanism of the prophylactic effects of atrial natriuretic peptide in an acute lung injury model. For the evaluation of the early phase of pulmonary inflammation, in vitro and in vivo studies using lipopolysaccharide were used. In the in vitro study, the effects of atrial natriuretic peptide on the induction of E-selectin by lipopolysaccharide in human pulmonary artery endothelial cells were evaluated. In the in vivo study, the effects of atrial natriuretic peptide on lipopolysaccharide-induced inflammatory cell infiltration and cytokine levels including tumor necrosis factor-alpha and interleukin-6 in the bronchoalveolar lavage fluid in the lungs of C57/B6 mice were examined. The number of myeloperoxidase-positive staining cells in the tissue sections of the lung of lipopolysaccharide-administered C57/B6 mice was also evaluated. Atrial natriuretic peptide significantly attenuated the up-regulation of E-selectin expression induced by lipopolysaccharide in human pulmonary artery endothelial cells. There were significantly lower cell counts and levels of tumor necrosis factor-alpha and interleukin-6 in the bronchoalveolar lavage fluid of atrial natriuretic peptide-treated mice compared to control mice after lipopolysaccharide injection. In addition, there were significantly fewer myeloperoxidase-positive cells in atrial natriuretic peptide-treated mice than in control mice after lipopolysaccharide injection. Atrial natriuretic peptide had a protective effect in the lipopolysaccharide-induced acute lung injury model. Atrial natriuretic peptide may be of value in therapeutic strategies aimed at the treatment of acute lung injury such as pneumonia or acute respiratory distress syndrome.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis