Merck
Dietary exposure of Nigerians to mutagens and estrogen-like chemicals.
International journal of environmental research and public health (2014-08-26)
Iyekhoetin Matthew Omoruyi, Derek Ahamioje, Raimo Pohjanvirta
ABSTRACT
Food and drinking water are poorly delineated sources of human exposure to chemical food mutagens and endocrine-disrupting chemicals. In this study, we investigated the presence of mutagens and chemicals exhibiting estrogenic activity in the daily diet of Nigerians, using in vitro assays. Commercially processed foods or snacks and various brands of pure water sachets were extracted by solid-phase extraction and liquid-liquid extraction, respectively. Mutagenicity was determined by the conventional Ames test and two complementary assays on two strains of Salmonella (TA 100 and TA 98), while the estrogenic activity was assessed by a yeast bioluminescent assay, using two recombinant yeast strains (Saccharomyces cerevisiae BMAEREluc/ERα and S. cerevisiae BMA64/luc). A third of the food varieties investigated (chin-chin, hamburger, suya and bean cake) were mutagenic in all three assays, either in the presence or absence of S9 mix. Of the packed water samples, five out of the sixteen investigated (31%), were found to be estrogenic, with estradiol and bisphenol A equivalents ranging from 0.79 to 44.0 ng/L and 124.2 to 1,000.8 ng/L, respectively. Hence, although the current situation in Nigeria does not appear to be substantially worse than, e.g., in Europe, regular monitoring is warranted in the future.
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O