Merck
  • Synthesis of novel tetrahydrobenzazepine derivatives and their cytoprotective effect on human lymphocytes.
Synthesis of novel tetrahydrobenzazepine derivatives and their cytoprotective effect on human lymphocytes.
Archiv der Pharmazie (2015-02-11)
Milena R Simic, Miroslava Stankovic, Boris M Mandic, Vele V Tesevic, Vladimir M Savic
ABSTRACT
Cytoprotective compounds such as amifostine play an important role in chemo- and radiotherapy due to their ability to reduce the side effects of these treatments. Our work was initiated with the intention to design, synthesise and test a new class of heterocyclic compounds that would have an antioxidative profile with the potential to be further developed as cytoprotective agents. The design was based on the privileged tetrahydrobenzazepine scaffold found in many natural products with a wide range of biological properties. This structure was further functionalised with moieties known to possess antioxidative features such as tertiary amine and styrene double bond. A series of eight tetrahydrobenzazepine derivatives of isoquinoline, 3,4-dihydro-β-carboline and pyridine were synthesised employing the Heck reaction as a key transformation. Some of the prepared compounds were tested for their in vitro effects on chromosome aberrations in peripheral human lymphocytes using the cytochalasin-B blocked micronucleus (MN) assay. Three tetrahydrobenzoazepine derivatives showed significant cytoprotective properties, comparable or even better to those of the radioprotective agent amifostine.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Supelco
Diethyl ether, analytical standard
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF