Merck
  • Identification, quantification, and sensory characterization of steviol glycosides from differently processed Stevia rebaudiana commercial extracts.
Identification, quantification, and sensory characterization of steviol glycosides from differently processed Stevia rebaudiana commercial extracts.
Journal of agricultural and food chemistry (2014-11-14)
María Inés Espinoza, Jean-Paul Vincken, Mark Sanders, Cristian Castro, Markus Stieger, Eduardo Agosin
ABSTRACT
Stevia rebaudiana is known for its sweet-tasting ent-kaurene diterpenoid glycosides. Several manufacturing strategies are currently employed to obtain Stevia sweeteners with the lowest possible off-flavors. The chemical composition of four commercial S. rebaudiana extracts, obtained by different technologies, was characterized using UHPLC-ESI-MS(n). The composition of one of the ethanol-crystallized extracts (EC2) was entirely rebaudioside A, whereas the enzymatically modified (EM) extract contained the lowest concentration of this compound (2.7 mg/100 mg). The membrane-purified (MP) extract had the highest content of minor natural steviol glycosides (23.7 mg/100 mg total extract) versus an average of 2.4 mg/100 mg total extract for the EC samples. Thirteen trained panelists evaluated sweetness, bitterness, licorice, and metallic attributes of all four extracts. The highest licorice intensity (p ≤ 0.05) was found for MP. Both samples EC1 and EC2, despite their different chemical compositions, showed no significant differences in sensory perception.
Product Description
Supelco
Sucralose, analytical standard
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sucralose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucralose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucralose, United States Pharmacopeia (USP) Reference Standard
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sucrose, European Pharmacopoeia (EP) Reference Standard
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture