Merck
  • N-acetylcysteine relieves oxidative stress and protects hippocampus of rat from radiation-induced apoptosis by inhibiting caspase-3.
N-acetylcysteine relieves oxidative stress and protects hippocampus of rat from radiation-induced apoptosis by inhibiting caspase-3.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-03-18)
Jianguo Li, Zhuangzhi Meng, Guowei Zhang, Yonghua Xing, Li Feng, Saijun Fan, Feiyue Fan, Batu Buren, Qiang Liu
ABSTRACT
It has been recognized that radiation-induced effects remain a significant risk. An accumulation of reactive oxygen species (ROS) is considered to be one factor that contributes to neurodegenerative changes. The aim of our study was to investigate the potential radioprotective effects of NAC. Male Sprague-Dawley rats underwent radiation. Irradiation was performed at room temperature with a 4-Gy dose of radiation. A dose of N-acetylcysteine (NAC) was performed 15 min prior to irradiation intraperitoneally. The methods of immunohistochemistry, TUNEL staining, Nissl staining, qRT-PCR, analysis of reactive oxygen species and Western blot were performed. In conclusion, our results demonstrate that NAC inhibits apoptosis induced by irradiation via the inhibition of caspase-3. We demonstrated a decrease in caspase-3 mRNA that was present at 24h of NAC treatment. Such mRNA decrease was accompanied by a decrease of protein. In the present study, NAC effectively antagonized oxidation induced by irradiation. These results provide evidence that the neural protective effect and the antioxidant effect of NAC contribute to metabolic activity.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, ≥90% (HPLC), powder
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)