Merck
  • Investigation of nonylphenol and nonylphenol ethoxylates in sewage sludge samples from a metropolitan wastewater treatment plant in Turkey.
Investigation of nonylphenol and nonylphenol ethoxylates in sewage sludge samples from a metropolitan wastewater treatment plant in Turkey.
Talanta (2014-10-05)
Seçil Ömeroğlu, Fadime Kara Murdoch, F Dilek Sanin
ABSTRACT
Nonylphenol ethoxylates (NPEOs) have drawn significant attention within the last decade for both scientific and legislative reasons. In Turkey, the Regulation Regarding the Use of Domestic and Urban Sludges on Land states a limit value for the sum of nonylphenol (NP), nonylphenol monoethoxylate (NP1EO) and nonylphenol diethoxylate (NP2EO) as NPE (NPE=NP+NP1EO+NP2EO). Unfortunately a standard method for the determination of these chemicals has not been yet set by the authorities and no data exists about the concentrations of NP and NPEOs in sewage sludge in Turkey. The aim of this study is to propose simple and easily applicable extraction and measurement techniques for 4-n-nonylphenol (4-n-NP), NP, NP1EO and NP2EO in sewage sludge samples and investigate the year round concentrations in a Metropolitan Wastewater Treatment Plant (WWTP) in Turkey. Different extraction techniques and GC/MS methods for sewage sludge were tested. The best extraction method for these compounds was found to be ultrasonication (5 min) using acetone as the solvent with acceptable recovery of analytes suggested by USEPA and other studies. The optimized extraction method showed good repeatability with relative standard deviations (RSDs) less than 6%. The recovery of analytes were within acceptable limits suggested by USEPA and other studies. The limits of detection (LODs) were 6 µg kg(-1) for NP and NP1EO, 12 µg kg(-1) for NP2EO and 0.03 µg kg(-1) for 4-n-NP. The developed method was applied to sewage sludge samples obtained from the Central WWTP in Ankara, Turkey. The sum NPE (NP+NP1EO+NP2EO) was found to be in between 5.5 µg kg(-1) and 19.5 µg kg(-1), values which are in compliance with Turkish and European regulations.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Supelco
Cyclohexane, analytical standard
Supelco
Methanol, analytical standard
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material