Merck
  • Altered pulmonary artery endothelial-smooth muscle cell interactions in experimental congenital diaphragmatic hernia.
Altered pulmonary artery endothelial-smooth muscle cell interactions in experimental congenital diaphragmatic hernia.
Pediatric research (2015-01-13)
Shannon N Acker, Gregory J Seedorf, Steven H Abman, Eva Nozik-Grayck, Katherine Kuhn, David A Partrick, Jason Gien
ABSTRACT
Pulmonary hypertension (PH) secondary to vascular remodeling contributes to poor outcomes in congenital diaphragmatic hernia (CDH), however mechanisms responsible are unknown. We hypothesized that pulmonary artery endothelial cell (PAEC) dysfunction contributes to smooth muscle cell (SMC) hyperplasia in experimental CDH. PAEC and SMC were isolated from fetal sheep with experimental CDH and controls. SMC growth was assessed alone and with SOD plus catalase and during coculture with control or CDH PAEC with and without ET-1 siRNA transfection. ET-1 protein was measured in PAEC and SMC lysates and supernatant. ROS production was measured in normal and CDH PAECs with and without ET-1 siRNA. PAEC growth and tube formation were measured with SOD plus catalase. CDH SMC growth was decreased and increased with coculture with CDH PAEC more than control PAEC. Treatment of CDH PAEC with SOD plus catalase or ET-1 siRNA prevented the increase in SMC growth seen with coculture. ET-1 protein was increased in CDH PAEC and SMC. ROS production was increased in CDH PAEC and decreased with ET-1 SiRNA. SOD plus catalase restored CDH PAEC growth and tube formation. PAEC dysfunction in experimental CDH increases SMC proliferation via ET-1 induced ROS production by PAEC.
Product Description
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Ethanolamine, analytical standard
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human EDN1
Supelco
Aucubin, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Ethanolamine, ≥99%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Aucubin, primary reference standard
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized