Merck
  • Signaling mechanism underlying the promotion of keratinocyte migration by angiotensin II.
Signaling mechanism underlying the promotion of keratinocyte migration by angiotensin II.
Molecular pharmacology (2014-12-05)
Hiroki Sakai, Kenji Matsuura, Yoshie Tanaka, Takeshi Honda, Teruo Nishida, Makoto Inui
ABSTRACT
Re-epithelialization begins early during skin wound healing and is regulated by various growth factors and cytokines. Angiotensin II promotes the migration of keratinocytes and thereby contributes to wound healing. We investigated the mechanism by which angiotensin II stimulates human keratinocyte migration. Angiotensin II-induced keratinocyte migration was inhibited by an angiotensin II type 1 receptor (AT1R) antagonist (candesartan) or an angiotensin II type 2 receptor (AT2R) antagonist (PD123319) as well as by depletion of AT1R or AT2R. A biased agonist for AT1R, [Sar(1),Ile(4),Ile(8)]angiotensin II, induced cell migration, whereas depletion of β-arrestin2 inhibited angiotensin II-induced migration. Angiotensin II-induced migration was blocked by neutralizing antibodies to transforming growth factor-β (TGF-β) as well as by the TGF-β receptor inhibitor SB431542. The amount of TGF-β1 was increased in the culture medium of angiotensin II-treated cells, and this effect was inhibited by candesartan or PD123319. Both angiotensin II- and TGF-β-induced cell migration were inhibited by neutralizing antibodies to the epidermal growth factor (EGF) receptor but not by those to EGF receptor ligands. Angiotensin II-induced phosphorylation of the EGF receptor, and this effect was inhibited by candesartan, PD123319, SB431542, or depletion of β-arrestin2, but not by neutralizing antibodies to heparin-binding EGF-like growth factor. Our results indicate that β-arrestin-dependent signaling downstream of AT1R as well as AT2R signaling are necessary for angiotensin II-induced keratinocyte migration, and that such signaling promotes generation of the active form of TGF-β, consequent activation of the TGF-β receptor, and transactivation of the EGF receptor by the TGF-β receptor.
Product Description
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture