Merck
  • Greater frequency of CD5-negative CD8(+) T cells against human immunodeficiency virus type 1 than other viruses is consistent with adaptation to antigenic variation.
Greater frequency of CD5-negative CD8(+) T cells against human immunodeficiency virus type 1 than other viruses is consistent with adaptation to antigenic variation.
AIDS research and therapy (2014-09-23)
Stephen J Penney, Maureen E Gallant, Michael D Grant
ABSTRACT
The CD5 protein antagonizes phosphorylation events downstream of T cell receptor (TCR) engagement to decrease T cell responsiveness. CD5-negative T cell clones respond preferentially over their CD5(+) counterparts against cells with low human histocompatibility-linked leukocyte antigen (HLA) levels. In human immunodeficiency virus type 1 (HIV-1) infection, CD5(-)CD8(+) T cells increase in prevalence with disease progression. To investigate potential causes of this expansion of CD5(-)CD8(+) T cells in HIV-1 infection, we compared CD5 expression on CD8(+) T cells reactive against HIV-1 peptides, common viral peptides and a self peptide that together span a broad range of TCR avidities in the context of the common HLA-A2 class I restriction molecule. Following stimulation, CD5 expression on peptide-specific CD8(+) T cells was assessed by flow cytometry. In healthy controls, there was no significant difference in the CD5(+) percentage of CD8(+) T cells specific for common viral peptides, but a lower percentage of those responding against a common self peptide expressed CD5. The same relationship occurred in HIV-infected individuals, however, a lower percentage of HIV peptide-specific CD8(+) T cells than other viral peptide-specific CD8(+) T cells expressed CD5. In terms of overall CD5 expression level at the peptide-specific responder population level, HIV-specific CD8(+) T cells resembled those responsive against the self peptide, despite much higher avidity TCR/HLA/peptide interactions. This deficit in CD5 expression selective for HIV-specific CD8(+) T cells is consistent with in vivo adaptation to low avidity HIV peptide variants and has potential consequences for CD8(+) T cell expansion, cross-reactivity and autoreactivity.
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Interleukin-7 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-7 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
SAFC
HEPES
SAFC
HEPES
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
IL-7 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source