Merck
Determination of selenium and its compounds in marine organisms.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS) (2014-12-04)
Małgorzata Anita Bryszewska, Amund Måge
ABSTRACT
In this study, we investigate the type and quantity of selenium compounds in fish and marine organisms, using ion-pair reversed phase LC–ICP-MS, developed and applied for the analysis of Atlantic cod, Atlantic salmon, Greenland halibut, Atlantic herring, blue mussel, common crab, scallop, calanus, and Euphasia super. Of the samples examined, the lowest level of selenium was found in farmed Atlantic salmon (0.17 mg Se kg(−1) dm). The total selenium extraction efficiency by phosphate buffer was 2.5 times higher in sea plankton and shellfish samples than in fish samples. Analysis of Se species in each hydrolysate obtained by proteolysis showed the presence of selenomethionine, which constituted 41.5% of the selenium compounds detected in hydrolysates of Atlantic herring and 98.4% of those in extracts of Atlantic salmon. Inorganic compounds, such as selenates and selenites, were detected mainly in sea plankton and shellfish samples (<0.13 mg Se kg(−1) wm), although no correlation was found between the presence of inorganic compounds and total selenium concentration. The accuracy of the total selenium determination was validated using a certified reference material (oyster tissue (NIST 1566b)). A lyophilised powder of cod (Gadus morhua) was used to validate speciation analysis, enzymatic hydrolysis of lyophilised powder of cod recovered 54 ± 6% of total selenium, and SeMet constituted 83.5 ± 5.28% of selenium detected in hydrolysates. The chromatographic detection limits were, respectively, 0.30 ng mL(−1), 0.43 ng mL(−1), 0.54 ng mL(−1), 0.55 ng mL(−1), 0.57 ng mL(−1) and 0.72 ng mL(−1) for selenate, selenomethionine, selenite, Se-methyl-selenocysteine, selenocystine and selenomethionine selenoxide.The data on selenium concentrations and speciation presented here could be useful in estimating levels of selenium intake by seafood consumption.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Supelco
Triethylamine, analytical standard
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Triethylamine, ≥99%