Merck
  • Role of STAT5 and epigenetics in lactation-associated upregulation of multidrug transporter ABCG2 in the mammary gland.
Role of STAT5 and epigenetics in lactation-associated upregulation of multidrug transporter ABCG2 in the mammary gland.
American journal of physiology. Endocrinology and metabolism (2014-08-15)
Alex Man Lai Wu, Mingdong Yang, Pooja Dalvi, Andrei L Turinsky, Wei Wang, Darci Butcher, Sean E Egan, Rosanna Weksberg, Patricia A Harper, Shinya Ito
ABSTRACT
The multidrug resistance efflux transporter ATP-binding cassette subfamily G member 2 (ABCG2) is not only overexpressed in certain drug-resistant cancers but is also highly expressed in the mammary gland during lactation, carrying xenobiotics and nutrients into milk. We sought to investigate the molecular mechanisms involved in the upregulation of ABCG2 during lactation. Expression profiling of different mouse Abcg2 mRNA isoforms (E1a, E1b, and E1c) revealed that E1b is predominantly expressed and induced in the lactating mouse mammary gland. Despite this induction, analyses of CpG methylation status and published ChIP-seq datasets reveal that E1b promoter sequences in the virgin gland are already hypomethylated and marked with the open chromatin histone mark H3K4me2. Using a forced-weaning model to shut down lactation, we found that within 24 h there was a significant reduction in Abcg2 mRNA expression and a loss of signal transducer and activator of transcription-5 (STAT5) occupancy at the mouse Abcg2 gene. Luciferase reporter assays further showed that some of these STAT5-binding regions that contained interferon-γ-activated sequence (GAS) motifs function as an enhancer after prolactin treatment. We conclude that Abcg2 is already poised for expression in the virgin mammary gland and that STAT5 plays an important role in Abcg2 expression during lactation.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
BIS-TRIS
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-STAT5A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
PIPES, BioXtra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
PIPES, ≥99% (titration)
SAFC
BIS-TRIS
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)