Merck
  • Magnetic nanoclusters engineered by polymer-controlled self-assembly for the accurate diagnosis of atherosclerotic plaques via magnetic resonance imaging.
Magnetic nanoclusters engineered by polymer-controlled self-assembly for the accurate diagnosis of atherosclerotic plaques via magnetic resonance imaging.
Macromolecular bioscience (2014-04-18)
Myeong-Hoon Kim, Bongjune Kim, Eun-Kyung Lim, Yuna Choi, Jihye Choi, Eunjung Kim, Eunji Jang, Hyo Seon Park, Jin-Suck Suh, Yong-Min Huh, Seungjoo Haam
ABSTRACT
Oleyl dextran-coated magnetic nanoclusters (ODMCs) are fabricated for the accurate detection of macrophage-rich atherosclerotic plaques using magnetic resonance (MR) imaging. Dextran is introduced to the cluster surface of magnetic nanocrystals (MNCs) through self-assembly using amphiphilic oleic acid-conjugated dextran (ODex) to provide not only hydrophilicity but also a high affinity to macrophages. Enhanced magnetism of the ODMCs is engineered by optimizing the degree of substitution (DS) of the oleyl group in ODex and the concentration of ODex used for the synthesis of ODMC. Consequently, ODMCs exhibit significantly increased T2 relaxivity and excellent macrophage-targeting ability without cytotoxicity, in vitro. Moreover, in vivo T2-weighted MR imaging after intravenous injection of ODMCs into a rat artery balloon injury model demonstrates considerable MR contrast strength efficacy in the plaques of the injured carotid artery. These novel ODMCs may offer a highly efficient MR imaging nanoprobes for the selective diagnosis of atherosclerosis.
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Phosphotungstic Acid Solution, 10 % (w/v)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Benzyl ether, ≥98%, FCC, FG
Sigma-Aldrich
Oleylamine, technical grade, 70%
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
DCC, puriss., ≥99.0% (GC)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Biebrich Scarlet-Acid Fuchsin Solution
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard