Merck
  • PET/CT with 18F-FDG- and 18F-FBEM-labeled leukocytes for metabolic activity and leukocyte recruitment monitoring in a mouse model of pulmonary fibrosis.
PET/CT with 18F-FDG- and 18F-FBEM-labeled leukocytes for metabolic activity and leukocyte recruitment monitoring in a mouse model of pulmonary fibrosis.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2014-12-30)
Benjamin Bondue, Félicie Sherer, Gaetan Van Simaeys, Gilles Doumont, Dominique Egrise, Yousof Yakoub, François Huaux, Marc Parmentier, Sandrine Rorive, Sébastien Sauvage, Simon Lacroix, Olivier Vosters, Paul De Vuyst, Serge Goldman
ABSTRACT
Idiopathic pulmonary fibrosis is characterized by a progressive and irreversible respiratory failure. Validated noninvasive methods able to assess disease activity are essential for prognostic purposes as well as for the evaluation of emerging antifibrotic treatments. C57BL/6 mice were used in a murine model of pulmonary fibrosis induced by an intratracheal instillation of bleomycin (control mice were instilled with a saline solution). At different times after instillation, PET/CT with (18)F-FDG- or (18)F-4-fluorobenzamido-N-ethylamino-maleimide ((18)F-FBEM)-labeled leukocytes was performed to assess metabolic activity and leukocyte recruitment, respectively. In bleomycin-treated mice, a higher metabolic activity was measured on (18)F-FDG PET/CT scans from day 7 to day 24 after instillation, with a peak of activity measured at day 14. Of note, lung mean standardized uptake values correlated with bleomycin doses, histologic score of fibrosis, lung hydroxyproline content, and weight loss. Moreover, during the inflammatory phase of the model (day 7), but not the fibrotic phase (day 23), bleomycin-treated mice presented with an enhanced leukocyte recruitment as assessed by (18)F-FBEM-labeled leukocyte PET/CT. Autoradiographic analysis of lung sections and CD45 immunostaining confirm the higher and early recruitment of leukocytes in bleomycin-treated mice, compared with control mice. (18)F-FDG- and (18)F-FBEM-labeled leukocyte PET/CT enable monitoring of metabolic activity and leukocyte recruitment in a mouse model of pulmonary fibrosis. Implications for preclinical evaluation of antifibrotic therapy are expected.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Xylazine, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
SAFC
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Bleomycin sulfate, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Fluoro-2-deoxy-D-glucose, glycosylation inhibitor, glucose analog
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
SKU
Pack Size
Availability
Price
Quantity
Tantalum(V) ethoxide, packaged for use in deposition systems