Merck
Application of the Fenton process in the elimination of helminth eggs.
Journal of water and health (2014-12-05)
Sandra Isabel Escobar-Megchún, Hugo Alejandro Nájera-Aguilar, Magaly González-Hilerio, Javier Gutiérrez-Jiménez, Rubén Fernando Gutiérrez-Hernández, María Neftalí Rojas-Valencia
ABSTRACT
This study relates to a method for evaluating the degradation efficiency of Ascaris lumbricoides eggs through a Fenton reaction, using hydrogen peroxide (H2O2) at 100, 250, and 500 mg/L, ferrous sulfate (FeSO4) at 458 mg/L Fe(2+) and pH values of 3, 4.5, and 6. The experiments were conducted according to a 3(2) experimental design, with 1:1, 3:1, and 5:1 (H2O2/Fe(2+)) molar ratios. The oxidation and flocculation stages were performed at 130 rpm during 2 hours and at 25-30 rpm during 20 min, respectively. As a result of the Fenton reaction, an average of 91.2% Ascaris lumbricoides egg degradation was achieved at pH 6 using a 500 mg/L dose of H2O2 and a 3:1 (H2O2/Fe(2+)) molar ratio. Thus, this process is an alternative for eliminating parasites that are resistant to conventional disinfection processes and significantly reduces the human health hazard they represent.
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1m, diameter 0.5mm, as drawn, 99.998%
Iron, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, rod, 500mm, diameter 12.7mm, as drawn, 99.95%
Iron, foil, 50x50mm, thickness 2mm, as rolled, 99.99+%
Iron, tube, 200mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, rod, 200mm, diameter 25mm, as drawn, 98+%
Iron, tube, 100mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%
Iron, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 300x300mm, thickness 2.0mm, as rolled, armco« soft ingot 99.8%
Iron, wire reel, 1m, diameter 0.025mm, as drawn, 99.99+%