Merck
  • Evaluation on activity of cytochrome p450 enzymes in turbot via a probe drug cocktail.
Evaluation on activity of cytochrome p450 enzymes in turbot via a probe drug cocktail.
Journal of aquatic animal health (2014-11-05)
Zhi-Qiang Chang, Jian Li, Qian-Qian Zhai
ABSTRACT
Cytochrome P450s (CYPs) are the main catalytic enzymes for metabolism by a variety of endogenous and exogenous substrates in mammals, fish, insects, etc. We evaluated the application of a multidrug cocktail on changes in CYP1, CYP2, and CYP3 activity in Turbot Scophthalmus maximus. The probe drugs were a combination of caffeine (5 mg/kg body weight), dapsone (5 mg/kg), and chlorzoxazone (10 mg/kg). After a single intraperitoneal injection of the cocktail, the concentration of all three probe drugs in the plasma increased quickly to a peak and then decreased gradually over 24 h. Pharmacokinetic profiles of the three probe drugs were determined using a noncompartmental analysis, and the typical parameters were calculated. In the assay for CYP induction, pretreatment with rifampicin significantly reduced the typical pharmacokinetic metrics for caffeine and chlorzoxazone, but not dapsone, indicating that the activity of CYP1 and CYP2 in turbot were induced by rifampicin.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-Aminophenyl sulfone, 97%
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Dapsone, VETRANAL®, analytical standard
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Chlorzoxazone
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, BioXtra
Dapsone, European Pharmacopoeia (EP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Caffeine, European Pharmacopoeia (EP) Reference Standard
Rifampicin, European Pharmacopoeia (EP) Reference Standard