Merck
  • Atrial natriuretic peptide protects against cisplatin-induced acute kidney injury.
Atrial natriuretic peptide protects against cisplatin-induced acute kidney injury.
Cancer chemotherapy and pharmacology (2014-11-12)
Takashi Nojiri, Hiroshi Hosoda, Toru Kimura, Koichi Miura, Shin Ishikane, Takeshi Tokudome, Yasushi Shintani, Masayoshi Inoue, Mikiya Miyazato, Meinoshin Okumura, Kenji Kangawa
ABSTRACT
Cisplatin is an effective chemotherapeutic agent used in the treatment of a wide variety of malignancies. Acute kidney injury (AKI) is the major toxicity associated with cisplatin and sometimes necessitates a reduction in dose or discontinuation of treatment. Atrial natriuretic peptide (ANP) is secreted by the heart and exerts a wide range of renoprotective effects, including anti-inflammatory activity. The objective of this study was to investigate the protective effects of ANP on cisplatin-induced AKI in mice. Mice were randomly divided into three groups: control, cisplatin (20 mg/kg, intraperitoneal)/vehicle treatment, and cisplatin/ANP (1.5 μg/kg/min via osmotic-pump, subcutaneous) treatment. At 72 h after cisplatin injection, serum blood urea nitrogen and creatinine, urine albumin/creatinine, and renal expression of mRNAs encoding tumor necrosis factor-α, interleukin (IL)-1β, IL-6, intercellular adhesion molecule-1, monocyte chemoattractant protein-1, and transforming growth factor (TGF)-β were measured using real-time polymerase chain reaction. Histological changes were also evaluated. ANP treatment significantly attenuated cisplatin-induced increases in serum blood urea nitrogen and creatinine, urine albumin/creatinine, and renal expression of IL-1β, IL-6, intercellular adhesion molecule-1, and monocyte chemoattractant protein-1 mRNAs. Cisplatin-induced renal dysfunction and renal tubular necrosis were thus attenuated by ANP treatment. Our results indicate that ANP exhibits a protective effect against cisplatin-induced AKI in mice. ANP may thus be of value in prophylactic strategies aimed at mitigating the adverse effects associated with chemotherapy agents, including cisplatin.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Cisplatin, European Pharmacopoeia (EP) Reference Standard