Merck
  • Degradation of sulphamethazine by means of an improved photo-Fenton process involving a hydrogen peroxide systematic dosage.
Degradation of sulphamethazine by means of an improved photo-Fenton process involving a hydrogen peroxide systematic dosage.
Environmental technology (2014-06-25)
Evelyn Yamal-Turbay, Lydia Pérez González, Moisès Graells, Montserrat Pérez-Moya
ABSTRACT
Despite being acknowledged as an emerging contaminant, sulphamethazine (SMT) degradation has received scarce attention in the advanced oxidation processes field. Thus, this work addresses the degradation of SMT in water solutions (12 L of 25mgL-1 samples) by means of a photo-Fenton process and a systematic H202 dosage protocol that enhances its performance. A conventional photo-Fenton process led to 86% mineralization after 120 min treatment when adding the Fenton reactants at once (initial concentrations were 10mgL-1 Fe(II) and 200mgL-1 H2O2). Conversely, the process achieved the total mineralization of the samples in less than 75 min when the same amount of H202 was continuously dosed according to a conveniently tuned dosage protocol. In both cases, total SMT degradation was achieved within 10 min. Hence, this work's aim is to determine the efficient dosage conditions of H2O2. The results show that a significant improvement of the photo-Fenton mineralization of SMT solutions is possible by adjusting the dosage of H2O2.
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Iron, IRMM®, certified reference material, 0.5 mm wire
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Iron, foil, 25mm disks, thickness 0.007mm, 99.85%
Iron, foil, 10mm disks, thickness 0.15mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.038mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.015mm, 99.99+%
Iron, foil, 25mm disks, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 2m coil, thickness 0.125mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Iron, foil, 25mm disks, thickness 0.015mm, 99.85%
Iron, foil, 4mm disks, thickness 0.004mm, 99.99+%
Iron, foil, 1m coil, thickness 0.025mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.1mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.008mm, 99.85%
Iron, foil, 25mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.01mm, 99.99+%
Iron, foil, 2m coil, thickness 0.15mm, hard, 99.5%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%