Merck
  • Expression of cereblon protein assessed by immunohistochemicalstaining in myeloma cells is associated with superior response of thalidomide- and lenalidomide-based treatment, but not bortezomib-based treatment, in patients with multiple myeloma.
Expression of cereblon protein assessed by immunohistochemicalstaining in myeloma cells is associated with superior response of thalidomide- and lenalidomide-based treatment, but not bortezomib-based treatment, in patients with multiple myeloma.
Annals of hematology (2014-04-02)
Shang-Yi Huang, Chung-Wu Lin, Hsiu-Hsia Lin, Ming Yao, Jih-Luh Tang, Shang-Ju Wu, Yao-Chang Chen, Hsiao-Yun Lu, Hsin-An Hou, Chien-Yuan Chen, Wen-Chien Chou, Woei Tsay, Sheng-Je Chou, Hwei-Fang Tien
ABSTRACT
Cereblon (CRBN) is essential for the anti-myeloma (MM) activity of immunomodulatory drugs (IMiDs), such as thalidomide and lenalidomide. However, the clinical implications of CRBN in MM patients are unclear. Using immunohistochemical (IHC) staining on paraffin-embedded bone marrow sections, the expression of CRBN protein in myeloma cells (MCs) was assessed in 40 relapsed/refractory MM (RRMM) patients who received lenalidomide/dexamethasone (LD) and 45 and 22 newly diagnosed MM (NDMM) patients who received thalidomide/dexamethasone (TD) and melphalan/bortezomib/prednisolone (MVP), respectively. IHC staining were scored on a scale representing the diffuseness and intensity of positive-staining MCs (range, 0-8) and a score ≥4.5 was used for CRBN positivity (CRBN(+)) on a cut-point analysis of all possible scores and response of TD and LD. Compared to CRBN(+) NDMM patients, CRBN(-) NDMM patients had more international staging system (ISS) III (26 vs. 61 %, respectively; P = 0.006). In the LD and TD cohorts, the response rate (RR) was higher in CRBN(+) patients than CRBN(-) patients (LD 79 vs. 33 %, respectively; P = 0.005) (TD 75 vs. 29 %, respectively; P = 0.005); however, this trend was not observed in the MVP cohort. In the LD and TD cohorts, the positive and negative prediction value of CRBN(+) for treatment response was 79 and 67 % and 75 and 71 %, respectively. Multivariate analysis showed that CRBN(+) was a significant factor associated with superior RR for LD and TD. The data suggest that expression of CRBN protein in MCs assessed using the IHC is a feasible approach to predict the response of IMiDs in MM patients.
Product Description
Supelco
Prednisolone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Prednisolone, ≥99%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Prednisolone, VETRANAL®, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Prednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
Prednisolone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Melphalan, powder
Sigma-Aldrich
(±)-Thalidomide, ≥98%, powder
Prednisolone for peak identification, European Pharmacopoeia (EP) Reference Standard