Merck
  • [Caffeine--common ingredient in a diet and its influence on human health].
[Caffeine--common ingredient in a diet and its influence on human health].
Roczniki Panstwowego Zakladu Higieny (2012-08-30)
Regina Wierzejska
ABSTRACT
Caffeine is widely consumed by people of all ages. In the last period a market of caffeine-containing products, particularly energy drinks and food supplements increased. Caffeine for years is under discussion, whether has positive whether adverse impact on health. Children are a group of special anxieties. Caffeine is a stimulant of central nervous system and therefore is probably the most commonly used psychoactive substance in the world. The physiological effect of caffeine and the lack of nutrition value causes a great interest its impact on health, especially with reference to the risk of cardiovascular diseases. Results of scientific research are not clear. The influence of caffeine on the human body is conditioned with the individual metabolism of caffeine which also depends on many endogenic and environmental factors. According to the current knowledge moderate caffeine intake by healthy adults at a dose level of 400 mg a day is not associated with adverse effects, but it also depends on other health determinants of a lifestyle. Excessive caffeine consumption can cause negative health consequences such as psychomotor agitation, insomnia, headache, gastrointestinal complaints. Adverse effect of caffeine intoxication is classified in World Health Organization's International Classification of Diseases (ICD-10). Metabolism of caffeine by pregnant woman is slowed down. Caffeine and its metabolites pass freely across the placenta into a fetus. For this reason pregnant women should limit caffeine intake. Children and adolescents should also limit daily caffeine consumption. It results from the influence of caffeine on the central nervous system in the period of rapid growth and the final stage of brain development, calcium balance and sleep duration. Average daily caffeine consumption in European countries ranging from 280-490 mg. The highest caffeine intake is in Scandinavian countries what results from the great consumption of the coffee. As far as caffeine consumption by Polish population is concerned there is very few data in this subject so far. In the nineties of the previous century it was 141 mg per day, whereas according to recent survey daily caffeine intake by women from the Warsaw region was 251 mg and 15% of examined women consumed an excessive quantity of caffeine (> or = 400 mg). Smokers consume more caffeine than nonsmokers, similarly to persons with mental illnesses. With reference to the caffeine consumption it should be underline that caffeine content in coffee and tea beverages varies greatly depending on the method of brewing whereas the content of caffeine in many brands of energy drinks can much vary. This should be taken into account in the daily caffeine intake.
Product Description
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard