Merck
  • Shaping of mesoporous AISBA-15 as cylindrical nano reactor for tertiary butylation of phenol.
Shaping of mesoporous AISBA-15 as cylindrical nano reactor for tertiary butylation of phenol.
Journal of nanoscience and nanotechnology (2014-04-18)
Govindasamy Chandrasekar, Martin Hartmann, Velayutham Murugesan
ABSTRACT
Mesoporous AISBA-15 with different n(Si)/n(Al) ratios (45, 136 and 215) in the powder (P) form were synthesized by hydrothermal technique. The powder materials were shaped into cylindrical extrudates (Ex) by compounding with additives such as bentonite (binder) and methylcellulose (plasticizer). The AISBA-15 materials were characterized by XRD, N2 adsorption, AAS, 27Al MAS-NMR and thermogravimetric analysis. The orderly growth of AISBA-15 is evidenced by its XRD patterns. The surface area and pore volume of AISBA-15 catalysts were around 950 m2/g and 1.3 cm3/g respectively. A decrease in surface area and pore volume were observed for AISBA-15 (Ex) materials in comparison to AISBA-15 (P) materials. This may be due to partial blocking of pore entry and surface area coverage of additives during the shaping of extrudates. Vapor phase alkylation of phenol with tert-butanol was carried out over AISBA-15 (P) and AISBA-15 (Ex) catalysts and the results revealed that both powder and extrudate samples showed similar activity for phenol conversion. The activity of AISBA-15 catalysts follows the order: AISBA-15 (45) > AISBA-15 (136) > AISBA-15 (215) for both powder and extrudate forms. The major products were 2-tert-butylphenol (2-TBP), 4-tert-butylphenol (4-TBP) and 2,4-di-tert-butylphenol (2,4-DTBP). The selectivity to para product is much higher than other products. The catalyst exhibited steady conversion, and deactivation by coke formation of the catalyst is much reduced in AISBA-15 materials.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Carbon, tube, graphite, 150mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, tube, graphite, 150mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon, tube, graphite, 50mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%
Carbon, foil, 150x150mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 150x150mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 100x100mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%
Carbon, foil, 150x150mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%
Carbon, foil, 15x15mm, thickness 2.0mm, hOpg
Carbon, foil, 150x150mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 200x200mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 25x25mm, thickness 0.5mm, pyrolytic graphite, 99.99%
Carbon, foil, 500x500mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, rod, 300mm, diameter 6.35mm, graphite, 100%
Carbon, foil, 250x250mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 500x500mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, rod, 100mm, diameter 50mm, graphite, 99.95%
Carbon, microleaf, 25x70mm, thinness 2.5μm, specific density 500μg/cm2, 99.997%
Carbon, tube, graphite, 150mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%