Skip to Content
Merck
All Photos(3)

Key Documents

C3014

Sigma-Aldrich

Activated charcoal

greener alternative

untreated, granular, 20-60 mesh

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€44.80
2.5 KG
€143.00

€44.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€44.80
2.5 KG
€143.00

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
SB.52

€44.80


Check Cart for Availability

Request a Bulk Order

vapor pressure

<0.1 mmHg ( 20 °C)

form

untreated, granular

autoignition temp.

842 °F

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

1375 μΩ-cm, 20°C (graphite)

particle size

20-60 mesh

mp

3550 °C (lit.)

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated charcoal (activated carbon) has been used:
  • As an adsorbent for p-cresol in the fermentation broth, during its synthesis from p-hydroxyphenylacetate catalyzed by p-hydroxyphenylacetate decarboxylase.[1]
  • To form a charcoal filter in a Drierite cartridge for removing organic compounds from the air for the odor delivery system.[2]
  • As an organic sorbent to study adsorption equilibrium of ammonia gas.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin C Daly et al.
PloS one, 8(11), e81863-e81863 (2013-11-28)
Sensory systems sample the external world actively, within the context of self-motion induced disturbances. Mammals sample olfactory cues within the context of respiratory cycles and have adapted to process olfactory information within the time frame of a single sniff cycle.
In-situ product recovery as a strategy to increase product yield and mitigate product toxicity.
Ng YL and Kuek YY
Open Biotechnology Journal, 7, 15-22 (2013)
Adsorption equilibria of ammonia gas on inorganic and organic sorbents at 298.15 K.
Helminen J, et al.
Journal of Chemical and Engineering Data, 46(2), 391-399 (2001)
Kohsuke Ohmatsu et al.
Nature chemistry, 6(1), 47-51 (2013-12-19)
The development of a general catalytic method for the direct and stereoselective construction of contiguous all-carbon quaternary stereocentres remains a formidable challenge in chemical synthesis. Here, we report a highly enantio- and diastereoselective [3+2] annulation reaction of 5-vinyloxazolidinones and activated
Nathalie Butt et al.
Ecological applications : a publication of the Ecological Society of America, 23(4), 936-943 (2013-07-20)
A typical way to quantify aboveground carbon in forests is to measure tree diameters and use species-specific allometric equations to estimate biomass and carbon stocks. Using "citizen scientists" to collect data that are usually time-consuming and labor-intensive can play a

Questions

1–2 of 2 Questions  
  1. What is the source of this activated carbon? Is it coconut based; coal based; lignite based, etc.?

    1 answer
    1. This item is chemically manufactured from raw materials of plant origin. The species is not identified and may vary.

      Helpful?

  2. I would like to know the surface area and the pore size of this product?

    1 answer
    1. Unfortunately, the pore size, pore volume and surface area of each lot of Product C3014 are not determined.
      A surface area of at least 1000 square meters per gram is typical for activated charcoal/activated carbon.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service