Merck
  • Template-free hydrothermal synthesis different morphologies of visible-light-driven BiVO4 photocatalysts.
Template-free hydrothermal synthesis different morphologies of visible-light-driven BiVO4 photocatalysts.
Journal of nanoscience and nanotechnology (2014-04-18)
Cai Xu, Gangqiang Zhu, Junlin Wu, Jia Liang
ABSTRACT
Monoclinic BiVO4 nano- and microstructures with a diversity of well-defined morphologies, such as nanoplates, dendrite leaves-like structures, sub-microrods, and microflowers were synthesized via a template-free hydrothermal process with bismuth nitrate and ammonium metavanadate as metal source. The crystal structures, morphologies and optical properties of the as-prepared samples were characterized by X-ray powder diffraction (XRD), transmission electron microscope (TEM), scanning electron microscopy (SEM), X-ray photoelectron spectroscopy (XPS) and UV-visible absorption spectra (UV-vis). Results showed that the pH value of the solution and the volume of ethylenediamine have great effect on the formation of these unique structures. The photocatalytic activity of these as-prepared samples had been tested by degradation of methylene blue under visible light, indicating that showed good photocatalytic performance.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, suitable for ion chromatography
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Methylene Blue solution, for microscopy
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Bismuth, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.97%