Merck
  • Technical and economic feasibility of polyester dyeing wastewater treatment by coagulation/flocculation and Fenton's oxidation.
Technical and economic feasibility of polyester dyeing wastewater treatment by coagulation/flocculation and Fenton's oxidation.
Environmental technology (2014-04-08)
Carmen S D Rodrigues, Rui A R Boaventura, Luis M Madeira
ABSTRACT
This study aims to investigate the efficiency of individual and integrated processes applied to organic matter abatement and biodegradability improvement of a polyester dyeing wastewater, namely coagulation/flocculation combined with Fenton's reagent (Approach 1), Fenton oxidation alone (Approach 2) and its integration with coagulation/flocculation (Approach 3). The effects of Fe2+ dose, initial concentration of the oxidant (H202) and temperature during Fenton's oxidation were evaluated in Approaches 1 and 2, whereas in Approach 3 the influence ofpH and flocculant dose was also assessed, during the coagulation/flocculation stage. Toxicity and biodegradability of the final effluent were also evaluated. After oxidation, a slight increase in the specific oxygen uptake rate of the effluent was observed (from 27.0 up to 28.5-30.0mg O2/(gVSSh)) and the inhibition to Vibrio fischeri was eliminated. An effluent that complies with discharge standards was obtained in all cases; however, Approach 3 revealed to be a promising solution for treating this effluent as it leads to smaller operating costs. Therefore, the use of dissolved iron resulting from Fenton's oxidation as coagulant in the second stage was shown to be an innovative, efficient and economically attractive strategy for treating these effluents.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Iron, IRMM®, certified reference material, 0.5 mm wire
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Iron, wire reel, 20m, diameter 0.25mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.075mm, as rolled, 99.99+%
Iron, foil, 15mm disks, thickness 0.1mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.5mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.075mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.05mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.003mm, 99.85%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.038mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, 10mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.0125mm, 99.85%
Iron, foil, 25mm disks, thickness 0.004mm, 99.85%
Iron, foil, 15mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 1m coil, thickness 0.20mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.038mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.005mm, 99.85%
Iron, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%