Merck
  • The role of NAD(+)-dependent isocitrate dehydrogenase 3 subunit α in AFB1 induced liver lesion.
The role of NAD(+)-dependent isocitrate dehydrogenase 3 subunit α in AFB1 induced liver lesion.
Toxicology letters (2013-11-12)
Chi Yang, Jue Fan, Zhenhong Zhuang, Yi Fang, Yanfeng Zhang, Shihua Wang
ABSTRACT
Aflatoxin B1 (AFB1) is a potent hepatocarcinogen that causes carcinogenesis in many animal species. In previous study, we found that isocitrate dehydrogenase 3α subunit (IDH3α) was upregulated in AFB1-induced carcinogenesis process. In this study, the sequences of IDH3α from various species were compared and the protein expression levels in different organs were examined, and the results showed that IDH3α was a widely distributed protein and shared highly conserved sequence in various species. In the same time, IDH3α was demonstrated to accumulate in a dose-dependent manner induced by AFB1 in cells, and was also up-regulated in the process of AFB1-induced liver lesion. Similar results were observed when H2O2 was used to replace AFB1. Over-expression of IDH3α increased the phosphorylation level of Akt (Protein kinase B) and neutralized the cellular toxicity induced by AFB1 or H2O2 and apoptosis induced by AFB1, while the reduced expression of IDH3α by siRNA decreased the phosphorylation, indicating that IDH3α played important roles in oxidative stress-induced PI3K/Akt pathway. Overall, the results suggested that AFB1 treatment could increase the expression of IDH3α, and the activated PI3K/Akt pathway by IDH3α eventually neutralized the apoptosis induced by AFB1.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Aflatoxin B1 solution, 3.79 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B1 solution, 3 μg/mL in benzene:acetonitrile (98:2), analytical standard
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Supelco
Aflatoxin B1 solution, 2 μg/mL in acetonitrile, analytical standard