Merck
  • Uptake of advanced glycation end products by proximal tubule epithelial cells via macropinocytosis.
Uptake of advanced glycation end products by proximal tubule epithelial cells via macropinocytosis.
Biochimica et biophysica acta (2013-06-12)
Marisa A Gallicchio, Leon A Bach
ABSTRACT
Chronic hyperglycaemia during diabetes leads to non-enzymatic glycation of proteins to form advanced glycation end products (AGEs) that contribute to nephropathy. We describe AGE uptake in LLC-PK1 and HK2 proximal tubule cell lines by macropinocytosis, a non-specific, endocytic mechanism. AGE-BSA induced dorsal circular actin ruffles and amiloride-sensitive dextran-TRITC uptake, significantly increased AGE-BSA-FITC uptake (167±20% vs BSA control, p<0.01) and was ezrin-dependent. AGE-BSA-FITC uptake was significantly inhibited by amiloride and inhibitors of Arf6, Rac1, racGEF Tiam1, PAK1 and actin polymerisation. AGE-BSA-FITC, Arf6 and PIP2 co-localised within dorsal circular actin ruffles. AGE-BSA increased PAK1 kinase activity (212±41% vs control, p<0.05) and protein levels of Tiam1, a Rac1 activator. AGE-BSA significantly increased TGF-β1 protein levels (160±6%, p<0.001 vs BSA), which were significantly inhibited by inhibitors of Arf6 (82±19%, p<0.001 vs AGE) and actin polymerisation (107±11%, p<0.001 vs AGE), suggesting AGEs partially exert their profibrotic effects via macropinocytosis. PAK1 and PIP5Kγ siRNA significantly decreased AGE-BSA-FITC uptake (81±6% and 64±7%, respectively, p<0.05 vs control for both), and AGE-stimulated TGF-β1 protein release (99±15% and 49±8% of control, p<0.05 and p<0.001, respectively). Inhibition of AGE uptake by macropinocytosis inhibitors and a neutralising TGF-β antibody, reversed the AGE-induced decrease in surface Na(+)K(+)ATPase, suggesting AGE uptake by macropinocytosis may contribute to diabetic kidney fibrosis and/or EMT by modulating this pump. Understanding methods of cellular uptake and signalling by AGEs may lead to novel therapies for diabetic nephropathy.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 670,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 1,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 12,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Supelco
Dextran, analytical standard, Set Mp 1,000-400,000
Supelco
Dextran, analytical standard, 5,000
Supelco
Dextran, analytical standard, 25,000
Supelco
Dextran, analytical standard, 410,000
Supelco
Dextran, analytical standard, 150,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Supelco
Dextran, analytical standard, 670,000