Merck
  • Serum 7 alpha-hydroxy-4-cholesten-3-one and selenohomocholyltaurine (SeHCAT) whole body retention in the assessment of bile acid induced diarrhoea.
Serum 7 alpha-hydroxy-4-cholesten-3-one and selenohomocholyltaurine (SeHCAT) whole body retention in the assessment of bile acid induced diarrhoea.
European journal of gastroenterology & hepatology (1996-02-01)
W G Brydon, H Nyhlin, M A Eastwood, M V Merrick
ABSTRACT
To assess the reliability of serum 7 alpha-hydroxy-4-cholesten-3-one (7 alpha-3ox-C) in the differential diagnosis of bile acid induced diarrhoea by comparison with 75selenohomocholyltaurine whole body retention (SeHCAT WBR). One hundred and sixty-four patients with chronic diarrhoea were investigated prospectively in two centres (Edinburgh and Sweden) by two different tests which measure bile acid loss or synthesis: the SeHCAT test which measures the 7-day SeHCAT WBR and serum 7 alpha-3ox-C which reflects the rate of bile acid synthesis. Forty-six patients had SeHCAT WBR of less than 10% (19 with ileal disease or resection, nine with idiopathic bile acid induced diarrhoea and 18 with miscellaneous causes for bile acid induced diarrhoea). All patients with ileal or idiopathic disease showed a favorable response to treatment as did 13 of the miscellaneous group. Serum 7 alpha-3ox-C was raised in all subjects with ileal disease/resection, seven patients with idiopathic disease and all subjects in the miscellaneous group who responded to treatment. Sixteen out of 118 patients with SeHCAT WBR greater than or equal to 10% had raised serum 7 alpha-3ox-C. The positive predictive value of serum 7 alpha-3ox-C was 74%. The high negative predictive value (98%) of serum 7 alpha-3ox-C indicates the possible use of this test for excluding bile acid malabsorption in this population. All but two subjects who responded to treatment had raised serum 7 alpha-3ox-C concentrations. The possibility that the sensitivity of the test can be improved by repeat testing needs to be further investigated. There was a significant correlation between fractional catabolic rate (FCR) SeHCAT and serum 7 alpha-3ox-C (r = 0.63, P < 0.0001). Further data are required to validate the reference range in women over 70 years of age.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Supelco
Acetone, analytical standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®