Merck
  • Comparison of serum and urinary levels of modified nucleoside, 1-methyladenosine, in cancer patients using a monoclonal antibody-based inhibition ELISA.
Comparison of serum and urinary levels of modified nucleoside, 1-methyladenosine, in cancer patients using a monoclonal antibody-based inhibition ELISA.
The Tohoku journal of experimental medicine (1995-05-01)
S Ishiwata, K Itoh, T Yamaguchi, N Ishida, M Mizugaki
ABSTRACT
To evaluate the clinical usefulness of serum 1-methyladenosine, several modifications have been made in our previously established inhibition ELISA system. Horseradish peroxidase (HRP) labeled anti-mouse IgG and 3,3',5,5'-tetramethylbenzidine (TMBZ) were used as a secondary antibody and a substrate, respectively. The second blocking was done just before the addition of the secondary antibody. The standard curve of the modified ELISA system showed good linearity between 1 and 1,000 ng/ml, and the detection limit was 50 pg/well. Using the ultrafiltrated-serum samples, serum 1-methyladenosine levels in healthy individuals and cancer patients were determined. The mean level of 1-methyladenosine in 31 healthy individuals was 28.3 +/- 7.9 ng/ml, and cut off value was set at 44.1 ng/ml (Mean+ 2SD). In cancer patients, elevated levels of serum 1-methyladenosine above the cut off value were detected in 4 out of 25 cases tested, though 11 cases had elevated urinary 1-methyladenosine levels above the cut off value (3.23 nmol/mumol creatinine). Since 1-methyladenosine has no interaction with serum proteins and its molecular weight is quite low, it might be rapidly excreted into the urine.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
SAFC
Glycine
SAFC
L-Tyrosine
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)