Merck
  • Influence of silver doping on electron transport in thin films of PbSe nanocrystals.
Influence of silver doping on electron transport in thin films of PbSe nanocrystals.
Advanced materials (Deerfield Beach, Fla.) (2012-11-20)
Moon Sung Kang, Ayaskanta Sahu, C Daniel Frisbie, David J Norris
ABSTRACT
Field-effect transistors are fabricated from thin films of Ag-doped PbSe nanocrystals to analyze the influence of electronically active impurities on electrical transport in this important material for nanocrystal applications. Data is collected as a function of nanocrystal size, dopant concentration, and temperature. Changes in the Fermi level and transport parameters indicate that Ag is acting as a p-type dopant (acceptor).
Sigma-Aldrich
Lead(II) selenide, 99.99%