Merck
  • Response surface methodology and Taguchi approach to assess the combined effect of formulation factors on minocycline delivery from collagen sponges.
Response surface methodology and Taguchi approach to assess the combined effect of formulation factors on minocycline delivery from collagen sponges.
Die Pharmazie (2013-06-28)
M V Ghica, M G Albu, L Popa, St Moisescu
ABSTRACT
An important aspect to be considered in the healing of acute or chronic cutaneous wounds is the associated potential infection management. Collagen, the most abundant protein of the extracellular matrix, with proven properties in wounds healing and tissues regeneration, is one of the most widely used biopolymers as carrier matrix for controlled drug delivery systems. For this reason, the purpose of the current paper is the development of some minocycline-loaded collagen topical sponges uncross-linked and cross-linked with glutaraldehyde, obtained by lyophilization of appropriate hydrogels prepared according to the 3-factor, 3-level face-centered central composite design. The determination of drug delivery from the sponges was performed by assessment of some physicochemical parameters involved in this complex process: sponges surface wettability, swelling ratio and the percentage of minocycline released from the sponges. The application of the response surface methodology allowed the setting of the formulation parameters optimum ranges, which ensure an adequate minocycline release to the application site. The design robustness was checked using the signal-to-noise ratio performance indicator. The optimum collagen-minocycline sponges determined based on the statistical screening technique could be suitable for topical drug delivery in infected wounds healing with moderate to high exudate.
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Minocycline hydrochloride, powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance