Merck
  • An RP-HPLC-UV method with SPE for cefotaxime in all-in-one total parenteral nutritional admixtures: application to stability studies.
An RP-HPLC-UV method with SPE for cefotaxime in all-in-one total parenteral nutritional admixtures: application to stability studies.
Journal of AOAC International (2013-06-19)
Muhammad Shahid Iqbal, Mohd Baidi Bahari, Yusrida Darwis, Muhammad Zahid Iqbal, Amer Hayat, Gantala Venkatesh
ABSTRACT
A simple and selective RP-HPLC-UV method with SPE was developed and validated for the quantification of cefotaxime in all-in-one total parenteral nutrition (AIO-TPN) admixtures. Chromatographic separation was achieved on a 5 pm particle size C18 DB column (250 x 4.6 mm id) using the mobile phase ammonium acetate (25 mM, pH 4.0)-50% acetonitrile in methanol (80 + 20, v/v). The flow rate was 0.9 mL/min and the detection wavelength was 254 nm. The analyte was extracted from AIO-TPN admixtures by means of an SPE method. The cefotaxime calibration curve was linear over a concentration range of 100-1400 microg/mL with a correlation coefficient of > or = 0.9994. The intraday accuracy and precision for cefotaxime were < or = -3.15 and < or = 3.08%, respectively, whereas the interday accuracy and precision were < or = -2.48 and < or = 2.25%, respectively. The method was successfully applied to stability studies of cefotaxime in the presence of micronutrients together with low and high concentrations of macronutrients in AIO-TPN admixtures. Cefotaxime was degraded by 13.00 and 26.05% at room temperature (25 +/- 2 degrees C) after 72 h in low and high macronutrient concentration formulations of AIO-TPN admixtures, respectively. The values of cefotaxime degradation rates for low and high macronutrient concentration formulations of AIO-TPN admixtures were -0.164 and -0.353, respectively. These results indicated that there was a higher rate of degradation in the AIO-TPN admixture formulations containing high concentrations of macronutrients.
Product Description
Cefotaxime acid, European Pharmacopoeia (EP) Reference Standard
Cefotaxime sodium, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Cefotaxime sodium salt, suitable for plant cell culture, BioReagent, powder or crystals
Sigma-Aldrich
Cefotaxime sodium salt, potency: 916-964 μg per mg
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Caffeine, European Pharmacopoeia (EP) Reference Standard