Merck
Inhibitory effect of anethole in nonimmune acute inflammation.
Naunyn-Schmiedeberg's archives of pharmacology (2012-12-20)
Talita Perdigão Domiciano, Márcia Machado de Oliveira Dalalio, Expedito Leite Silva, Alessandra Mileni Versuti Ritter, Camila Fernanda Estevão-Silva, Fernando Seara Ramos, Silvana Martins Caparroz-Assef, Roberto Kenji Nakamura Cuman, Ciomar Aparecida Bersani-Amado
ABSTRACT
Anethole [1-methoxy-4-(1-propenyl)benzene] occurs naturally as a major component of the essential oil of star anise (Illicium verum Hook.f., family Illiciaceae), comprising more than 90 % of its volatile components. Studies showed that this substance has antioxidant, antibacterial, antifungal, and anesthetic properties. In this study, the anti-inflammatory properties of anethole in animal models of nonimmune acute inflammation such as croton oil-induced ear edema and carrageenan-induced pleurisy were investigated. The investigated parameters were edema formation, leukocyte migration, and inflammatory mediators involved. Oral administration of anethole at a dose of 250 and 500 mg/kg reduced both the volume of pleural exudates and the number of migrated leukocytes. Levels of nitric oxide (NO) and prostaglandins (PGE2) in the inflammatory exudate were reduced by treatment with anethole, but levels of tumor necrosis factor-α and interleukin-1β were not significantly altered. In ear edema, the oral treatment with anethole inhibited the formation of exudate and the activity of myeloperoxidase, but not after topical administration. These results suggest that the anethole may be effective in controlling some nonimmune acute inflammation-related disease, probably by an inhibitory action on production and/or release of PGE2 and NO.
Product Description
Supelco
trans-Anethole, analytical standard
Sigma-Aldrich
trans-Anethole, ≥99%, FCC, FG
Sigma-Aldrich
trans-Anethole, 99%
Sigma-Aldrich
Croton oil
Supelco
trans-Anethole, Pharmaceutical Secondary Standard; Certified Reference Material