Merck
  • Design, synthesis and molecular docking of α,β-unsaturated cyclohexanone analogous of curcumin as potent EGFR inhibitors with antiproliferative activity.
Design, synthesis and molecular docking of α,β-unsaturated cyclohexanone analogous of curcumin as potent EGFR inhibitors with antiproliferative activity.
Bioorganic & medicinal chemistry (2012-12-19)
Yun-Yun Xu, Yi Cao, Hailkuo Ma, Huan-Qiu Li, Gui-Zhen Ao
ABSTRACT
A type of novel α,β-unsaturated cyclohexanone analogous, which designed based on the curcumin core structure, have been discovered as potential EGFR inhibitors. These compounds exhibit potent antiproliferative activity in two human tumor cell lines (Hep G2 and B16-F10). Among them, compounds I(3) and I(12) displayed the most potent EGFR inhibitory activity (IC(50) = 0.43 μM and 1.54 μM, respectively). Molecular docking of I(12) into EGFR TK active site was also performed. This inhibitor nicely fitting the active site might well explain its excellent inhibitory activity.
Sigma-Aldrich
Cyclohexanone, 99.8%
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Supelco
Cyclohexanone, Selectophore, for ion-selective electrodes, ≥99.5%
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Supelco
Cyclohexanone, analytical standard