Merck
  • Optimization of sample pretreatment methods for simultaneous determination of dolasetron and hydrodolasetron in human plasma by HPLC-ESI-MS.
Optimization of sample pretreatment methods for simultaneous determination of dolasetron and hydrodolasetron in human plasma by HPLC-ESI-MS.
Journal of chromatographic science (2012-05-31)
Yuming Hu, Shuo Chen, Jitao Chen, Guozhu Liu, Bo Chen, Shouzhuo Yao
ABSTRACT
A high-performance liquid chromatographic method coupled with electrospray mass spectrometry was developed for the simultaneous determination of dolasetron and its major metabolite, hydrodolasetron, in human plasma. A new sample pretreatment method, i.e., salt induced phase separation extraction (SIPSE), was proposed and compared with four other methods, i.e., albumin precipitation, liquid-liquid extraction, hydrophobic solvent-induced phase separation extraction and subzero-temperature induced phase separation extraction. Among these methods, SIPSE showed the highest extraction efficiency and the lowest matrix interferences. The extraction recoveries obtained from the SIPSE method were all more than 96% for dolasetron, hydrodolasetron and ondansetron (internal standard). The SIPSE method is also very fast and easy because protein precipitation, analyte extraction and sample cleanup are combined into one simple process by mixing acetonitrile with plasma and partitioning with 2 mol/L sodium carbonate aqueous solution. The correlation coefficients of the calibration curves were all more than 0.997, in the range of 7.9-4750.0 ng/mL and 4.8-2855.1 ng/mL for dolasetron and hydrodolasetron, respectively. The limits of quantification were 7.9 and 4.8 ng/mL for dolasetron and hydrodolasetron, respectively. The intra-day and inter-day repeatability were all less than 10%. The method was successfully applied to the pharmacokinetic study of dolasetron.
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Dolasetron mesylate hydrate, ≥98% (HPLC), powder