Merck
  • Caesium fluoride-promoted Stille coupling reaction: an efficient synthesis of 9Z-retinoic acid and its analogues using a practical building block.
Caesium fluoride-promoted Stille coupling reaction: an efficient synthesis of 9Z-retinoic acid and its analogues using a practical building block.
Chemical communications (Cambridge, England) (2008-12-03)
Takashi Okitsu, Kinya Iwatsuka, Akimori Wada
ABSTRACT
A highly efficient and rapid total synthesis of 9Z-retinoic acid was accomplished by caesium fluoride-promoted Stille coupling reaction; using a common building block, 9Z-retinoic acid analogues were also prepared by the same method without isomerisation of the Z-double bond.
Sigma-Aldrich
Cesium Fluoride ChemBeads, NSC 84270
Sigma-Aldrich
Cesium fluoride, 99%
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)