Merck
Dissolution nature of cesium fluoride by water molecules.
The journal of physical chemistry. B (2006-02-24)
Nongmaithem Jiten Singh, Hai-Bo Yi, Seung Kyu Min, Mina Park, Kwang S Kim
ABSTRACT
The structures, stabilities, thermodynamic quantities, dissociation energies, infrared spectra, and electronic properties of CsF hydrated by water molecules are investigated by using density functional theory, Møller-Plesset second-order perturbation theory (MP2), coupled cluster theory with singles, doubles, and perturbative triples excitations (CCSD(T)), and ab initio molecular dynamic (AIMD) simulations. It is revealed that at 0 K three water molecules (as a global minimum structure) begin to half-dissociate the Cs-F, and six water molecules (though not a global minimum energy structure) can dissociate it. By the combination of the accurate CCSD(T) conformational energies for Cs(H2O)6 at 0 K with the AIMD thermal energy contribution, it reveals that the half-dissociated structure is the most stable at 0 K, but this structure (which is still the most stable) changes to the dissociated structure above 50 K. The spectra of CsF(H2O)(1-6) from MP2 calculations and the power spectra of CsF(H2O)6 from 50 and 100 K AIMD simulations are also reported.
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99%
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Cesium Fluoride ChemBeads, NSC 84270