Merck
  • Development, characterization and efficacy of niosomal diallyl disulfide in treatment of disseminated murine candidiasis.
Development, characterization and efficacy of niosomal diallyl disulfide in treatment of disseminated murine candidiasis.
Nanomedicine : nanotechnology, biology, and medicine (2012-08-04)
Maroof Alam, Swaleha Zubair, Mohammad Farazuddin, Ejaj Ahmad, Arbab Khan, Qamar Zia, Abida Malik, Owais Mohammad
ABSTRACT
In the current study, a novel niosome based formulation of diallyl disulfide (DADS) was evaluated for its potential to treat disseminated candidiasis in mouse model. Among various non-ionic surfactants tested, niosome formulation prepared using Span 80 was found to be most efficient in the entrapment of DADS. The DADS loaded niosomes had size dimensions in the range of 140 ± 30 nm with zeta potential of -30.67 ± 4.5. Liver/kidney function tests as well as histopathologic studies suggested that noisome-based DADS formulations are safe at the dose investigated. When administered to Candida albicans infected animals, the DADS bearing niosomal formulation cleared the fungal burden and increased their survival much efficiently than its free form. In this study, a novel niosomal formulation of the antifungal DADS was utilized in a murine candidiasis model, resulting in more efficient fungal clearance compared to the free formulation.
Sigma-Aldrich
Span® 80, nonionic surfactant
Sigma-Aldrich
Allyl disulfide, technical grade, 80%
Supelco
Span® 80, suitable for GC
Supelco
Span® 80, viscosity 1000-2000 mPa.s (20 °C)
Sigma-Aldrich
Allyl disulfide, ≥80%, FG