Merck
  • UBE2D3 facilitates NHEJ by orchestrating ATM signalling through multi-level control of RNF168.
UBE2D3 facilitates NHEJ by orchestrating ATM signalling through multi-level control of RNF168.
Nature communications (2024-06-13)
Zeliha Yalçin, Shiu Yeung Lam, Marieke H Peuscher, Jaco van der Torre, Sha Zhu, Prasanna V Iyengar, Daniel Salas-Lloret, Inge de Krijger, Nathalie Moatti, Ruben van der Lugt, Mattia Falcone, Aurora Cerutti, Onno B Bleijerveld, Liesbeth Hoekman, Román González-Prieto, Jacqueline J L Jacobs
ABSTRACT
Maintenance of genome integrity requires tight control of DNA damage response (DDR) signalling and repair, with phosphorylation and ubiquitination representing key elements. How these events are coordinated to achieve productive DNA repair remains elusive. Here we identify the ubiquitin-conjugating enzyme UBE2D3 as a regulator of ATM kinase-induced DDR that promotes non-homologous end-joining (NHEJ) at telomeres. UBE2D3 contributes to DDR-induced chromatin ubiquitination and recruitment of the NHEJ-promoting factor 53BP1, both mediated by RNF168 upon ATM activation. Additionally, UBE2D3 promotes NHEJ by limiting RNF168 accumulation and facilitating ATM-mediated phosphorylation of KAP1-S824. Mechanistically, defective KAP1-S824 phosphorylation and telomeric NHEJ upon UBE2D3-deficiency are linked to RNF168 hyperaccumulation and aberrant PP2A phosphatase activity. Together, our results identify UBE2D3 as a multi-level regulator of NHEJ that orchestrates ATM and RNF168 activities. Moreover, they reveal a negative regulatory circuit in the DDR that is constrained by UBE2D3 and consists of RNF168- and phosphatase-mediated restriction of KAP1 phosphorylation.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-MSL3-like 1 Antibody, from rabbit
Sigma-Aldrich
Anti-PP2A Antibody, C subunit, clone 1D6, clone 1D6, Upstate®, from mouse
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Ubiquitinylated proteins Antibody, clone FK2, clone FK2, Upstate®, from mouse
Sigma-Aldrich
Anti-RNF168 Antibody, from rabbit, purified by affinity chromatography