Merck
An mTORC1-Dependent Mouse Model for Cardiac Sarcoidosis.
Journal of the American Heart Association (2023-09-26)
Carlos Bueno-Beti, Clarice X Lim, Alexandros Protonotarios, Petra Lujza Szabo, Joseph Westaby, Mario Mazic, Mary N Sheppard, Elijah Behr, Ouafa Hamza, Attila Kiss, Bruno K Podesser, Markus Hengstschläger, Thomas Weichhart, Angeliki Asimaki
ABSTRACT
Background Sarcoidosis is an inflammatory, granulomatous disease of unknown cause affecting multiple organs, including the heart. Untreated, unresolved granulomatous inflammation can lead to cardiac fibrosis, arrhythmias, and eventually heart failure. Here we characterize the cardiac phenotype of mice with chronic activation of mammalian target of rapamycin (mTOR) complex 1 signaling in myeloid cells known to cause spontaneous pulmonary sarcoid-like granulomas. Methods and Results The cardiac phenotype of mice with conditional deletion of the tuberous sclerosis 2 (TSC2) gene in CD11c+ cells (TSC2fl/flCD11c-Cre; termed TSC2KO) and controls (TSC2fl/fl) was determined by histological and immunological stains. Transthoracic echocardiography and invasive hemodynamic measurements were performed to assess myocardial function. TSC2KO animals were treated with either everolimus, an mTOR inhibitor, or Bay11-7082, a nuclear factor-kB inhibitor. Activation of mTOR signaling was evaluated on myocardial samples from sudden cardiac death victims with a postmortem diagnosis of cardiac sarcoidosis. Chronic activation of mTORC1 signaling in CD11c+ cells was sufficient to initiate progressive accumulation of granulomatous infiltrates in the heart, which was associated with increased fibrosis, impaired cardiac function, decreased plakoglobin expression, and abnormal connexin 43 distribution, a substrate for life-threatening arrhythmias. Mice treated with the mTOR inhibitor everolimus resolved granulomatous infiltrates, prevented fibrosis, and improved cardiac dysfunction. In line, activation of mTOR signaling in CD68+ macrophages was detected in the hearts of sudden cardiac death victims who suffered from cardiac sarcoidosis. Conclusions To our best knowledge this is the first animal model of cardiac sarcoidosis that recapitulates major pathological hallmarks of human disease. mTOR inhibition may be a therapeutic option for patients with cardiac sarcoidosis.
Sigma-Aldrich
BAY 11-7082, The BAY 11-7082, also referenced under CAS 19542-67-7, controls the biological activity of TNF-α. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Sigma-Aldrich
Anti-Pan Cadherin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Anti-Connexin-43 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution