Skip to Content
Merck
All Photos(1)

Documents

196870

Sigma-Aldrich

BAY 11-7082

≥98% (HPLC), solid, NF-kB/TNF-α inhibitor, Calbiochem

Synonym(s):

BAY 11-7082, (E)3-[(4-Methylphenyl)sulfonyl]-2-propenenitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2S
CAS Number:
Molecular Weight:
207.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

BAY 11-7082, The BAY 11-7082, also referenced under CAS 19542-67-7, controls the biological activity of TNF-α. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C10H9NO2S/c1-9-3-5-10(6-4-9)14(12,13)8-2-7-11/h2-6,8H,1H3/b8-2+

InChI key

DOEWDSDBFRHVAP-KRXBUXKQSA-N

General description

Potential anti-inflammatory agent that selectively and irreversibly inhibits the TNF-α-inducible phosphorylation of IκBα (IC50 = 10 µM), resulting in a decreased expression of NF-κB and of adhesion molecules. Does not affect constitutive IκBα autophosphorylation. Inhibits TNF-α-induced surface expression of the endothelial-leukocyte cell adhesion molecules E-selectin, VCAM-1, and ICAM-1. A 100 mM (10 mg/483 µl) solution of BAY 11-7082 (Cat. No. 196871) in DMSO is also available is also available.
Potential anti-inflammatory agent. Selectively and irreversibly inhibits the TNFα-inducible phosphorylation of IκB-α (IC50 = 10 µM) without affecting the constitutive IκB-α phosphorylation. Decreases nuclear translocation of NFκB and inhibits TNFα-induced surface expression of the endothelial-leukocyte cell adhesion molecules E-selectin, VCAM-1, and ICAM-1. E-selectin inhibition was shown to be irreversible (IC50 ~10 µM). Also reversibly increased the activity of p38 kinase and JNK-1 and stimulated tyrosine phosphorylation of a 130-140 kDa protein.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TNF-α-inducible phosphorylation of IκBα
Reversible: no
Target IC50: 10 µM inhibiting TNF-α-inducible phosphorylation of IκBα

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kamthong PJ. and Wu, M-C. 2001. Biochem. J. 356.
Izban, K.F., et al. 2000. Hum. Pathol.31, 1482.
Pierce, J.W., et al. 1997. J. Biol. Chem. 272, 21096.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhihong Chen et al.
The Journal of clinical investigation, 133(22) (2023-09-21)
Monocytes and monocyte-derived macrophages (MDMs) from blood circulation infiltrate glioblastoma (GBM) and promote growth. Here, we show that PDGFB-driven GBM cells induce the expression of the potent proinflammatory cytokine IL-1β in MDM, which engages IL-1R1 in tumor cells, activates the
Alejandro Sarrion-Perdigones et al.
Current protocols in molecular biology, 131(1), e122-e122 (2020-06-17)
Multiplex experimentation that can assay multiple cellular signaling pathways in the same cells requires orthogonal genetically encoded reporters that report over large dynamic ranges. Luciferases are cost-effective, versatile candidates whose output signals can be sensitively detected in a multiplex fashion.
Mohammed Soutto et al.
Cancer cell international, 21(1), 444-444 (2021-08-23)
H. pylori infection is the main risk factor for gastric cancer. In this study, we investigated H. pylori-mediated activation of STAT3 and NF-κB in gastric cancer, using in vitro and in vivo models. To investigate the activation of NF-κB and
Suji Kim et al.
Cells, 9(9) (2020-09-04)
Preeclampsia (PE) is a pregnancy-specific disorder associated with hypertension and proteinuria. Since there is no proven method to treat PE, early prediction and accurate diagnosis are essential for appropriate management of the disease. Thus, reliable biomarkers for diagnosing PE need
Yan Liu et al.
Journal of translational medicine, 20(1), 167-167 (2022-04-11)
Calcific aortic valve disease (CAVD) is the most commonly valvular disease in the western countries initiated by inflammation and abnormal calcium deposition. Currently, there is no clinical drug for CAVD. Neutrophil elastase (NE) plays a causal role in inflammation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service