Merck
  • Transplantable human motor networks as a neuron-directed strategy for spinal cord injury.
Transplantable human motor networks as a neuron-directed strategy for spinal cord injury.
iScience (2021-08-13)
Zachary T Olmsted, Cinzia Stigliano, Annalisa Scimemi, Tatiana Wolfe, Jose Cibelli, Philip J Horner, Janet L Paluh
ABSTRACT
To repair neural circuitry following spinal cord injury (SCI), neural stem cell (NSC) transplantation has held a primary focus; however, stochastic outcomes generate challenges driven in part by NSC differentiation and tumor formation. The recent ability to generate regionally specific neurons and their support cells now allows consideration of directed therapeutic approaches with pre-differentiated and networked spinal neural cells. Here, we form encapsulated, transplantable neuronal networks of regionally matched cervical spinal motor neurons, interneurons, and oligodendrocyte progenitor cells derived through trunk-biased neuromesodermal progenitors. We direct neurite formation in alginate-based neural ribbons to generate electrically active, synaptically connected networks, characterized by electrophysiology and calcium imaging before transplantation into rodent models of contused SCI for evaluation at 10-day and 6-week timepoints. The in vivo analyses demonstrate viability and retention of interconnected synaptic networks that readily integrate with the host parenchyma to advance goals of transplantable neural circuitry for SCI treatment.
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Brn-3a Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Synapsin I Antibody, serum, Chemicon®
Pricing and availability is not currently available.
SAFC
Insulin, Human Recombinant, for research or for further manufacturing use, dry powder
Pricing and availability is not currently available.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Pricing and availability is not currently available.