Merck
  • Comparison of coliforms and coliphages as tools for assessment of viral contamination in river water.
Comparison of coliforms and coliphages as tools for assessment of viral contamination in river water.
Applied and environmental microbiology (2004-06-09)
S Skraber, B Gassilloud, C Gantzer
ABSTRACT
The aim of the study was to evaluate the presence of pathogenic viruses in the Moselle River and to compare the usefulness of thermotolerant coliforms and somatic coliphages as tools for river water quality assessment in terms of viral contamination. Thermotolerant coliforms and somatic coliphages were enumerated by standardized methods in 170 samples of river water drawn from five sampling sites along the Moselle River (eastern France). BGM cell culture and integrated cell culture-reverse transcription-PCR DNA enzyme immunoassay were used to determine the presence of pathogenic viral genome (Enterovirus and Norovirus genogroup II [GGII]) and infectious Enterovirus spp. in 90 1-liter samples. No infectious Enterovirus spp. were isolated, but Enterovirus and Norovirus GGII genomes were detected in 38% of the samples. Norovirus GGII genome was mostly detected in winter, whereas Enterovirus genome was mostly detected in summer and fall. Somatic coliphages appeared to be less sensitive to higher river water temperature than thermotolerant coliforms. Furthermore, the number of river water samples positive for pathogenic viral genome increased with increasing concentration of somatic coliphages, whereas coliform concentration was unrelated to viral genome contamination. Consequently somatic coliphages, which are less sensitive to environmental factors than thermotolerant coliforms in river water, would provide a promising tool for assessment of river water quality in terms of fecal and viral pollution.
Product Description
Millipore
MCE Membrane Filter, 8.0 μm Pore Size, MF-Millipore, filter diam. 47 mm, hydrophilic