Skip to Content
Merck

Skip To

SCWP04700

MCE Membrane Filter, 8.0 μm Pore Size

MF-Millipore™, filter diam. 47 mm, hydrophilic

Sustainability features

Synonym(s):

Cellulose nitrate membrane filter discs, Hydrophilic MCE membrane filter cut discs, MF-Millipore™ Membrane Filter, 8 µm pore size, Mixed Cellulose Ester membrane filter discs

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
8.0 μm pore size, 84 % porosity
Material:
mixed cellulose esters (MCE) membrane, plain filter, white filter
Feature:
hydrophilic
Diam.:
47 mm
Sterility:
non-sterile
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore™, Millipore

sustainability

Sustainability features

parameter

620 mL/min-cm2 water flow rate, 65 L/min-cm2 air flow rate, 75 °C max. temp.

diam.

47 mm

filter L × W

1.9 cm × 4.2 cm

filter diam.

47 mm

thickness

135 μm

gravimetric extractables

6%

refractive index

n/D 1.515

matrix

MF-Millipore™

pore size

8.0 μm pore size, 84 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥0.42 bar, air with water at 23 °C

greener alternative category

shipped in

ambient

General description

Filter Code: SCWP
Need a custom membrane filter? Click here to request a custom membrane filter or unique and large package sizes that may not be available via our website.
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • QC of fluid holding tanks
  • Fluid monitoring
  • Air monitoring
  • Particle collection
  • Particle analysis

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCWP01300SSWP04700SCWP09025
description

MF-Millipore™, filter diam. 47 mm, hydrophilic

description

MF-Millipore™, filter diam. 13 mm, hydrophilic

description

MF-Millipore™, filter diam. 47 mm, hydrophilic

description

MF-Millipore™, filter diam. 90 mm, hydrophilic

diam.

47 mm

diam.

13 mm

diam.

47 mm

diam.

90 mm

pore size

8.0 μm pore size, 84 % porosity

pore size

8.0 μm pore size, 84 % porosity

pore size

3.0 μm pore size, 83 % porosity

pore size

84 % porosity, 8.0 μm pore size

manufacturer/tradename

MF-Millipore™

manufacturer/tradename

Millipore, MF-Millipore™

manufacturer/tradename

Millipore, MF-Millipore™

manufacturer/tradename

Millipore, MF-Millipore™

parameter

75 °C max. temp., 620 mL/min-cm2 water flow rate, 65 L/min-cm2 air flow rate

parameter

75 °C max. temp., 620 mL/min-cm2 water flow rate, 65 L/min-cm2 air flow rate

parameter

320 mL/min-cm2 water flow rate, 75 °C max. temp., 30 L/min-cm2 air flow rate

parameter

75 °C max. temp., 65 L/min-cm2 air flow rate, 620 mL/min-cm2 water flow rate

material

plain filter, white filter, mixed cellulose esters (MCE) membrane

material

plain filter, mixed cellulose esters (MCE) membrane, white filter

material

plain filter, white filter, mixed cellulose esters (MCE) membrane

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

sterility

non-sterile

sterility

-

sterility

non-sterile

sterility

non-sterile


Still not finding the right product?


Pictograms

Flame

Signal Word

Danger

Storage Class

4.1B - Flammable solid hazardous materials

Hazard Codes

Hazard Classifications

ED ENV 1 - Flam. Sol. 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

9036-19-5

CAS No.

9036-19-5

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters