Merck
  • Comparative assessment of toxic responses in 3D embryoid body differentiation model and mouse early embryos treated with 5-hydroxytryptophan.
Comparative assessment of toxic responses in 3D embryoid body differentiation model and mouse early embryos treated with 5-hydroxytryptophan.
Archives of toxicology (2020-09-15)
Olga Gordeeva, Andrey Gordeev
ABSTRACT
Pluripotent stem cells recapitulate in vitro the early developmental stages and are considered promising cell models for predictive developmental toxicity studies. To investigate the consistency between adverse drug effects on early development and the early stages of embryonic stem cell differentiation in three-dimensional (3D) in vitro culture, the toxic responses to 5-hydroxytryptophan (5-HTP; 0.5-2 mM) were evaluated in early mouse embryos and the embryoid body (EB) differentiation model. 3D architectures, developmental and differentiation dynamics and the cell death rates were analyzed in early mouse embryos (E2.5-E5.5) and EBs at 1 and 6 days of differentiation using a combination of confocal immunofluorescence microscopy with high content imaging analysis and quantitative gene expression analysis. Comparative analysis of toxic responses in early embryos and EBs revealed a similar dose- and stage-dependent decrease in the 5-HTP toxic effects during development and differentiation. The integral toxic responses in the early embryos and EBs were significantly dependent on their 3D architecture and cellular composition. Treatment with 5-HTP (1 mM and above) induced developmental arrest, growth inhibition, and increased cell death in the early embryos without the trophoblasts (E2.5) and those with impaired trophoblasts and in early EBs, whereas later embryos and EBs were more resistant due to the protection of the extraembryonic tissues. This study demonstrates that the EB differentiation model is a relevant 3D-model of early mammalian development and can be useful for the predictive evaluation of toxic and teratogenic effects in embryos at the preimplantation and early post-implantation developmental stages.
Sigma-Aldrich
Naphthol AS-BI phosphate, Technical grade, ≥98% (HPLC)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
5-Hydroxy-L-tryptophan, powder
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
M2 medium, With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Leukemia Inhibitory Factor from mouse, LIF, recombinant, expressed in E. coli, 10 μg/ml, buffered aqueous solution, suitable for cell culture