Merck
  • Phenoxy herbicides and fibrates potently inhibit the human chemosensory receptor subunit T1R3.
Phenoxy herbicides and fibrates potently inhibit the human chemosensory receptor subunit T1R3.
Journal of medicinal chemistry (2009-10-13)
Emeline L Maillet, Robert F Margolskee, Bedrich Mosinger
ABSTRACT
We show that phenoxyauxin herbicides and lipid-lowering fibrates inhibit human but not rodent T1R3. T1R3 as a coreceptor in taste cells responds to sweet compounds and amino acids; in endocrine cells of gut and pancreas T1R3 contributes to glucose sensing. Thus, certain effects of fibrates in treating hyperlipidemia and type II diabetes may be via actions on T1R3. Likewise, phenoxy herbicides may have adverse metabolic effects in humans that would have gone undetected in studies on rodents.
Product Description
Supelco
Dichloroprop-P, PESTANAL®, analytical standard
Supelco
1-Naphthaleneacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Indole-3-propionic acid, ≥99.0% (T)
Sigma-Aldrich
Indole-3-propionic acid, ≥97.0% (T)
Supelco
4-Chlorophenoxyacetic acid, PESTANAL®, analytical standard
Supelco
2,4-D, PESTANAL®, analytical standard
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
4-Chlorophenoxyacetic acid, BioReagent, suitable for plant cell culture, crystalline
Sigma-Aldrich
1-Naphthaleneacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Sigma-Aldrich
1-Naphthaleneacetic acid, technical grade
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
4-Chlorophenoxyacetic acid, ≥98.0% (T)
Supelco
Mecoprop, PESTANAL®, analytical standard
Supelco
Dichlorprop, PESTANAL®, analytical standard
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, 97%
Sigma-Aldrich
3-Indolepropionic acid, ReagentPlus®, 99%
Supelco
2-(2,4,5-Trichlorophenoxy)propionic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
2,3,5-Trichlorobenzoic acid, 97%
Supelco
Dicamba, PESTANAL®, analytical standard
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline
Sigma-Aldrich
1-Naphthylacetic acid, BioReagent, 1 mg/mL, suitable for plant cell culture
Sigma-Aldrich
Dicamba, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Indole-3-butyric acid, BioReagent, suitable for plant cell culture