Merck
  • Carbonic anhydrase activators: the first activation study of the human secretory isoform VI with amino acids and amines.
Carbonic anhydrase activators: the first activation study of the human secretory isoform VI with amino acids and amines.
Bioorganic & medicinal chemistry (2007-05-15)
Isao Nishimori, Saburo Onishi, Daniela Vullo, Alessio Innocenti, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
The secretory isozyme of human carbonic anhydrase (hCA, EC 4.2.1.1), hCA VI, has been cloned, expressed, and purified in a bacterial expression system. The kinetic parameters for the CO(2) hydration reaction proved hCA VI to possess a k(cat) of 3.4 x 10(5)s(-1) and k(cat)/K(M) of 4.9 x 10(7)M(-1)s(-1) (at pH 7.5 and 20 degrees C). hCA VI has a significant catalytic activity for the physiological reaction, of the same order of magnitude as the ubiquitous isoform CA I or the transmembrane, tumor-associated isozyme CA IX. A series of amino acids and amines were shown to act as CA VI activators, with variable efficacies. l-His, l-Trp, and dopamine showed weak CA VI activating effects (K(A)s in the range of 21-42 microM), whereas d-His, d-Phe, l-DOPA, l-Trp, serotonin, and some pyridyl-alkylamines were better activators, with K(A)s in the range of 13-19 microM. The best CA VI activators were l-Phe, d-DOPA, l-Tyr, 4-amino-l-Phe, and histamine, with K(A)s in the range of 1.23-9.31 microM. All these activators enhance k(cat), having no effect on K(M), participating thus in the rate determining step in the catalytic cycle, the proton transfer reactions between the enzyme active site and the environment.
Product Description
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
SAFC
L-Histidine
SAFC
L-Tyrosine
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
1-(2-Aminoethyl)piperazine, 99%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-Histidine, ≥98% (TLC)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%
Sigma-Aldrich
2-(2-Pyridyl)ethylamine, 95%
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
3,4-Dihydroxy-D-phenylalanine, powder, ≥95%
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland