Merck
  • Inflammation and DNA Methylation-Dependent Down-Regulation of miR-34b-5p Mediates c-MYC Expression and CRL4DCAF4 E3 Ligase Activity in Colitis-Associated Cancer.
Inflammation and DNA Methylation-Dependent Down-Regulation of miR-34b-5p Mediates c-MYC Expression and CRL4DCAF4 E3 Ligase Activity in Colitis-Associated Cancer.
The American journal of pathology (2020-01-24)
Chunmei Yang, Wenzhu Lu, Hongbo He, Hong Liu
ABSTRACT
miRNAs, a well-known group of noncoding RNAs, contribute to the pathogenesis of multiple diseases, including colitis-associated cancer (CAC). Our recent findings indicate that proinflammatory cytokines up-regulate c-MYC level, which subsequently activates cullin 4A and 4B (CUL4A/4B) and CRL4DCAF4 E3 ligases and promotes ubiquitination of suppression of tumorigenicity 7 in CAC. Herein, we identified and proved that miR-34b-5p can directly target c-MYC. In vitro oncogenic phenotype analyses and in vivo tumor formation assay indicated that miR-34b-5p overexpression could markedly decrease cell proliferation, colony formation, cell invasion, and tumor volumes. Overexpression of c-MYC in vitro could reverse the oncogenic phenotypes caused by miR-34b-5p up-regulation. In addition, the down-regulation of miR-34b-5p in CAC was dependent on the coregulation of the inflammatory microenvironment and DNA methylation. Collectively, our findings demonstrate that intracellular inflammation and DNA hypermethylation suppress miR-34b-5p expression, which limits its inhibitory effect on c-MYC and initiates the downstream events, including the induction of CRL4DCAF4 E3 ligase activity. The activated CRL4DCAF4 E3 ligase ubiquitinates suppression of tumorigenicity 7 and results in its degradation, eventually leading to the CAC tumorigenesis.
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Pricing and availability is not currently available.
Roche
DNA Isolation Kit for Cells and Tissues, sufficient for 10 isolation(s)
Pricing and availability is not currently available.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by ion-exchange chromatography, TLR ligand tested
Pricing and availability is not currently available.
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use
Pricing and availability is not currently available.
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.
Pricing and availability is not currently available.