Merck
Sphingosine-1-phosphate signaling and its role in disease.
Trends in cell biology (2011-10-18)
Michael Maceyka, Kuzhuvelil B Harikumar, Sheldon Milstien, Sarah Spiegel
ABSTRACT
The bioactive sphingolipid metabolite sphingosine-1-phosphate (S1P) is now recognized as a critical regulator of many physiological and pathophysiological processes, including cancer, atherosclerosis, diabetes and osteoporosis. S1P is produced in cells by two sphingosine kinase isoenzymes, SphK1 and SphK2. Many cells secrete S1P, which can then act in an autocrine or paracrine manner. Most of the known actions of S1P are mediated by a family of five specific G protein-coupled receptors. More recently, it was shown that S1P also has important intracellular targets involved in inflammation, cancer and Alzheimer's disease. This suggests that S1P actions are much more complex than previously thought, with important ramifications for development of therapeutics. This review highlights recent advances in our understanding of the mechanisms of action of S1P and its roles in disease.
Product Description
Avanti
Sphingosine-1-Phosphate (d20:1), Avanti Research - A Croda Brand 860662P, powder
Avanti
Sphingosine-1-Phosphate (d17:1), Avanti Research - A Croda Brand
Avanti
16:0-d31-18:1 PI, 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoinositol (ammonium salt), powder