Skip to Content
Merck
All Photos(2)

Key Documents

860042P

Avanti

16:0-d31-18:1 PI

1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoinositol (ammonium salt), powder

Synonym(s):

110923

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€231.00
500 μG
€918.00

€231.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€231.00
500 μG
€918.00

About This Item

Empirical Formula (Hill Notation):
C43H53NO13PD31
CAS Number:
Molecular Weight:
885.29
UNSPSC Code:
51191904
NACRES:
NA.25

€231.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (860042P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (860042P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 860042P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[NH4+].[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)OC[C@H](COP([O-])(=O)O[C@H]1C(O)C(O)C(O)[C@@H](O)C1O)OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C43H81O13P.H3N/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(45)55-35(34-54-57(51,52)56-43-41(49)39(47)38(46)40(48)42(43)50)33-53-36(44)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,35,38-43,46-50H,3-16,19-34H2,1-2H3,(H,51,52);1H3/b18-17-;/t35-,

InChI key

NPOZNLWSCICHRX-MPQXHUEUSA-N

General description

16:0-d31-18:1 PI or 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoinositol is a deuterated phospholipid. This lipid consists of a phosphatidylinositol group attached to palmitic acid and oleic acid.
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

16:0-d31-18:1 PI or 1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphoinositol has been used as an internal standard:
  • in yeast lipid profiling[1]
  • for the quantification of lipids using electrospray ionization tandem mass spectrometry (ESI-MS/MS)[2]
  • for the extraction of lipids from human bronchus tissue and epithelium samples using Bligh and Dyer method and for liquid chromatography-mass spectrometry (LC-MS) analysis [3]

Biochem/physiol Actions

Phosphatidylinositol (PI) is phosphorylated at their hydroxyls group and the phosphorylated PIs regulate several biological processes including membrane trafficking, recruitment of proteins and autophagy.[4]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (860042P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (860042P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Ter Braak et al.
Biochemical pharmacology, 81(5), 617-625 (2010-12-18)
Sphingosine-1-phosphate (S1P) acts as high affinity agonist at specific G-protein-coupled receptors, S1P(1-5), that play important roles e.g. in the cardiovascular and immune systems. A S1P receptor modulating drug, FTY720 (fingolimod), has been effective in phase III clinical trials for multiple
G van Echten-Deckert et al.
The Journal of biological chemistry, 272(25), 15825-15833 (1997-06-20)
The effect of six different structurally modified sphingosine analogues on biosynthesis of sphingolipids was studied in primary cultured murine cerebellar neurons. Treatment of cells with cis-4-methylsphingosine at micromolar levels resulted in a markedly decreased sphingolipid biosynthesis, whereas the other compounds
Nicole Zehethofer et al.
Chromatographia, 78(5-6), 403-413 (2015-03-10)
Airway epithelial cells play an important role in the pathogenesis of inflammatory lung diseases such as asthma, cystic fibrosis and COPD. Studies concerning the function of the lipid metabolism of the airway epithelium are so far based only on the
Nicholas J Blunsom et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(1), 158471-158471 (2019-06-08)
Phosphatidylinositol (PI) is a minor phospholipid with a characteristic fatty acid profile; it is highly enriched in stearic acid at the sn-1 position and arachidonic acid at the sn-2 position. PI is phosphorylated into seven specific derivatives, and individual species
Sarah Spiegel et al.
Nature reviews. Molecular cell biology, 4(5), 397-407 (2003-05-03)
The evolutionarily conserved actions of the sphingolipid metabolite, sphingosine-1-phosphate (S1P), in yeast, plants and mammals have shown that it has important functions. In higher eukaryotes, S1P is the ligand for a family of five G-protein-coupled receptors. These S1P receptors are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service