Merck
  • Development of a DIPG Orthotopic Model in Mice Using an Implantable Guide-Screw System.
Development of a DIPG Orthotopic Model in Mice Using an Implantable Guide-Screw System.
PloS one (2017-01-21)
Miguel Marigil, Naiara Martinez-Velez, Pablo D Domínguez, Miguel Angel Idoate, Enric Xipell, Ana Patiño-García, Marisol Gonzalez-Huarriz, Marc García-Moure, Marie-Pierre Junier, Hervé Chneiweiss, Elías El-Habr, Ricardo Diez-Valle, Sonia Tejada-Solís, Marta M Alonso
ABSTRACT
In this work we set to develop and to validate a new in vivo frameless orthotopic Diffuse Intrinsic Pontine Glioma (DIPG) model based in the implantation of a guide-screw system. It consisted of a guide-screw also called bolt, a Hamilton syringe with a 26-gauge needle and an insulin-like 15-gauge needle. The guide screw is 2.6 mm in length and harbors a 0.5 mm central hole which accepts the needle of the Hamilton syringe avoiding a theoretical displacement during insertion. The guide-screw is fixed on the mouse skull according to the coordinates: 1mm right to and 0.8 mm posterior to lambda. To reach the pons the Hamilton syringe is adjusted to a 6.5 mm depth using a cuff that serves as a stopper. This system allows delivering not only cells but also any kind of intratumoral chemotherapy, antibodies or gene/viral therapies. The guide-screw was successfully implanted in 10 immunodeficient mice and the animals were inoculated with DIPG human cell lines during the same anesthetic period. All the mice developed severe neurologic symptoms and had a median overall survival of 95 days ranging the time of death from 81 to 116 days. Histopathological analysis confirmed tumor into the pons in all animals confirming the validity of this model. Here we presented a reproducible and frameless DIPG model that allows for rapid evaluation of tumorigenicity and efficacy of chemotherapeutic or gene therapy products delivered intratumorally to the pons.
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Histone H3.3 Antibody, K27M mutant, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Nestin, Human Antibody, serum, from rabbit