![6-Fluoroindole 98%](/deepweb/assets/sigmaaldrich/product/structures/181/852/162b3c94-8bc6-4cb9-ac28-f2557e0ffe2d/640/162b3c94-8bc6-4cb9-ac28-f2557e0ffe2d.png)
6-Fluoroindole
![6-Fluoroindole 98%](/deepweb/assets/sigmaaldrich/product/structures/181/852/162b3c94-8bc6-4cb9-ac28-f2557e0ffe2d/640/162b3c94-8bc6-4cb9-ac28-f2557e0ffe2d.png)
Synonym(s):
NSC 520436
Empirical Formula (Hill Notation):
C8H6FN
CAS Number:
Molecular Weight:
135.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing